Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mypn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182992.2 | NP_892037.2 | 19 | 3998 | ||
GenBank mRNA | AK039718.1 | - | 3 | 1069 | ||
AK220521.1 | - | 19 | 3998 | |||
BC052872.1 | - | 5 | 666 |