Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167768.1 | NP_001161240.1 | 6 | 807 | ||
NM_172401.4 | NP_765989.3 | 6 | 826 | |||
GenBank mRNA | AK076460.1 | - | 4 | 492 | ||
AK079099.1 | - | 6 | 810 | |||
AK086717.1 | - | 5 | 623 | |||
AK162850.1 | - | 4 | 457 | |||
AK162914.1 | - | 6 | 788 | |||
AW049235.1 | - | 1 | 396 | |||
BC046986.1 | - | 5 | 745 | 6476504 |