Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026854.3 | NP_081130.1 | 3, 4 | 460 | ||
GenBank mRNA | AK004450.1 | - | 3, 4 | 435 | ||
AK144689.1 | - | 3, 4 | 435 | |||
BC075693.1 | - | 5, 6 | 554 | |||
BC139347.1 | - | 3, 4 | 426 | 8862369 | ||
BC139348.1 | - | 3, 4 | 426 | 8862370 |