Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026861.2 | NP_081137.2 | 8 | 1030 | ||
GenBank mRNA | AK035384.1 | - | 5 | 605 | ||
AK049109.1 | - | 8 | 979 | |||
AK158177.1 | - | 8 | 979 | |||
AK159755.1 | - | 3 | 388 | |||
BC017635.1 | - | 2 | 192 | |||
BC024467.1 | - | 8 | 1016 | 4975777 |