Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026861.2 | NP_081137.2 | 2 | 251 | ||
GenBank mRNA | AK049109.1 | - | 2 | 200 | ||
AK158177.1 | - | 2 | 200 | |||
AK182703.1 | - | 2 | 99 | |||
AK186783.1 | - | 2 | 99 | |||
BC024467.1 | - | 2 | 237 | 4975777 | ||
BY080555.1 | - | 2 | 251 |