Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026861.2 | NP_081137.2 | 7, 8 | 947 | ||
GenBank mRNA | AK035384.1 | - | 4, 5 | 522 | ||
AK049109.1 | - | 7, 8 | 896 | |||
AK158177.1 | - | 7, 8 | 896 | |||
AK159755.1 | - | 2, 3 | 305 | |||
BC017635.1 | - | 1, 2 | 109 | |||
BC024467.1 | - | 7, 8 | 933 | 4975777 |