Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175138.4 | NP_780347.2 | 4, 5 | 439 | ||
GenBank mRNA | AK030213.1 | - | 4, 5 | 373 | ||
AK047808.1 | - | 4, 5 | 440 | |||
BC138785.1 | - | 4, 5 | 384 | 8861807 | ||
BC138787.1 | - | 4, 5 | 384 | 8861809 |