Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haghl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271433.1 | NP_001258362.1 | 7 | 786 | ||
NM_001271434.1 | NP_001258363.1 | 7 | 786 | |||
NM_001271435.1 | NP_001258364.1 | 7 | 786 | |||
NM_001271436.1 | NP_001258365.1 | 6 | 600 | |||
NM_001271437.1 | NP_001258366.1 | 6 | 595 | |||
NM_026897.3 | NP_081173.1 | 6 | 771 | |||
GenBank mRNA | AK005274.1 | - | 6 | 767 | ||
AK012748.1 | - | 7 | 783 | |||
AK021220.1 | - | 6 | 602 | |||
AK032999.1 | - | 4 | 744 | |||
AK034798.1 | - | 7 | 761 | |||
AK047517.1 | - | 7 | 741 | |||
AK079632.1 | - | 4 | 935 | |||
AK080454.1 | - | 1 | 1722 | |||
AK154248.1 | - | 6 | 576 | |||
BC005586.1 | - | 7 | 732 | |||
BC030466.1 | - | 7 | 771 | 5253213 | ||
BC083322.1 | - | 7 | 725 | 6820885 |