Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102563.1 | NP_001096033.1 | 2 | 800 | ||
GenBank mRNA | AK158895.1 | - | 2 | 721 | ||
BF323287.1 | - | 1 | 80 | |||
CB246480.1 | - | 1 | 565 |
Gene Symbol: Pagr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC054744.1 | - | 565 |