Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026911.3 | NP_081187.2 | 4 | 657 | ||
GenBank mRNA | AI848281.1 | - | 1 | 362 | ||
AK007336.1 | - | 4 | 355 | |||
AK082928.1 | - | 4 | 356 | |||
AK088658.1 | - | 2 | 895 | |||
BC034270.2 | - | 4 | 532 |