Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026917.5 | NP_081193.2 | 2 | 335 | ||
GenBank mRNA | AB088044.1 | - | 1 | 101 | ||
AK007355.1 | - | 2 | 309 | |||
AK018755.1 | - | 2 | 301 | |||
AK046166.1 | - | 2 | 335 | |||
AK080541.1 | - | 2 | 301 | |||
AK169197.1 | - | 278 | ||||
AY166672.1 | - | 1 | 101 | |||
BC005531.1 | - | 2 | 310 | |||
BC018334.1 | - | 2 | 296 | |||
BC025126.1 | - | 2 | 320 | 3669723 | ||
BC052464.1 | - | 2 | 240 | 6834763 | ||
CJ181332.1 | - | 2 | 335 |