Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022885.2 | NP_075023.2 | 14 | 2162 | ||
GenBank mRNA | AF233321.1 | - | 7 | 1181 | ||
AF461761.1 | - | 14 | 2061 | |||
AK007424.1 | - | 5 | 682 | |||
AK051183.1 | - | 14 | 2162 | |||
AK088004.1 | - | 13 | 1906 | |||
AK090176.1 | - | 14 | 2071 | |||
AK166006.1 | - | 14 | 1972 | |||
AK187253.1 | - | 3 | 375 | |||
AK215365.1 | - | 1 | 118 | |||
BC029033.1 | - | 11 | 1604 | |||
BC029217.1 | - | 7 | 1014 | |||
BC033452.1 | - | 14 | 2061 | 3156622 |