Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133705.2 | NP_598466.1 | 3 | 345 | ||
GenBank mRNA | AK007535.1 | - | 3 | 327 | ||
AK150756.1 | - | 2 | 1356 | |||
AK152302.1 | - | 2 | 1356 | |||
AK165618.1 | - | 3 | 320 | |||
AK167622.1 | - | 3 | 301 | |||
AK178957.1 | - | 52 | ||||
BC006882.1 | - | 3 | 321 | 2647612 | ||
CJ154235.1 | - | 3 | 344 |