Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810010H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163473.1 | NP_001156945.1 | 3 | 257 | ||
GenBank mRNA | BC124997.1 | - | 2 | 211 | ||
BC125006.1 | - | 2 | 211 | |||
BY099414.1 | - | 3 | 257 | |||
CJ236692.1 | - | 3 | 227 |