Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133708.3 | NP_598469.1 | 4 | 243 | ||
GenBank mRNA | AK007463.1 | - | 4 | 234 | ||
AK144311.1 | - | 4 | 508 | |||
AK150473.1 | - | 5 | 379 | |||
AK182835.1 | - | 4 | 204 | |||
AK189202.1 | - | 4 | 203 | |||
AK189514.1 | - | 4 | 270 | |||
AK198797.1 | - | 4 | 203 | |||
AK206464.1 | - | 3 | 149 | |||
AK210576.1 | - | 4 | 262 | |||
AK215045.1 | - | 4 | 261 | |||
BC008116.1 | - | 4 | 202 | 3498021 | ||
BY063474.1 | - | 4 | 250 |