Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178027.4 | NP_821170.1 | 4 | 1124 | ||
GenBank mRNA | AK010160.1 | - | 4 | 1058 | ||
AK031589.1 | - | 4 | 1127 | |||
AK035321.1 | - | 4 | 1075 | |||
AK150929.1 | - | 4 | 1045 | |||
AK155578.1 | - | 4 | 1085 | |||
AK165539.1 | - | 4 | 1063 | |||
AK171154.1 | - | 4 | 1083 | |||
AK215961.1 | - | 2 | 169 | |||
BC029758.1 | - | 2 | 217 | |||
BC049180.1 | - | 4 | 1087 | 6314347 | ||
BC052721.1 | - | 4 | 1048 | 6809853 | ||
BC062972.1 | - | 4 | 1048 | 6809878 |