Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178027.4 | NP_821170.1 | 3 | 983 | ||
GenBank mRNA | AK010160.1 | - | 3 | 917 | ||
AK031589.1 | - | 3 | 986 | |||
AK150929.1 | - | 3 | 904 | |||
AK155578.1 | - | 3 | 944 | |||
AK165539.1 | - | 3 | 922 | |||
AK171154.1 | - | 3 | 942 | |||
AK215961.1 | - | 1 | 28 | |||
BC029758.1 | - | 1 | 76 | |||
BC049180.1 | - | 3 | 946 | 6314347 | ||
BC052721.1 | - | 3 | 907 | 6809853 | ||
BC062972.1 | - | 3 | 907 | 6809878 |