Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024134.2 | NP_001019305.1 | 2, 3 | 546 | ||
NM_001177872.1 | NP_001171343.1 | 2, 3 | 569 | |||
GenBank mRNA | AK007445.1 | - | 2, 3 | 570 | ||
BC027186.1 | - | 2, 3 | 546 | 4215801 |