Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026965.2 | NP_081241.1 | 3 | 329 | ||
GenBank mRNA | AK007659.1 | - | 3 | 327 | ||
AK054334.1 | - | 3 | 330 | |||
BC019467.1 | - | 3 | 313 | |||
BC049670.1 | - | 3 | 324 | 6535487 | ||
CT010388.1 | - | 3 | 291 |
Gene Symbol: A430057M04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046931.1 | - | 667 |