Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199188.1 | NP_001186117.1 | 3 | 328 | ||
NM_024225.5 | NP_077187.1 | 2 | 239 | |||
GenBank mRNA | AK007676.1 | - | 2 | 187 | ||
AK029463.1 | - | 2 | 199 | |||
AK031873.1 | - | 3 | 220 | |||
AK049129.1 | - | 3 | 220 | |||
AK078776.1 | - | 3 | 221 | |||
AK144710.1 | - | 3 | 221 | |||
AK152311.1 | - | 3 | 264 | |||
AK159904.1 | - | 2 | 144 | |||
AK167362.1 | - | 2 | 159 | |||
AK169545.1 | - | 2 | 460 | |||
AK169786.1 | - | 3 | 220 | |||
AK177366.1 | - | 1 | 52 | |||
AK184438.1 | - | 1 | 52 | |||
AK187575.1 | - | 1 | 52 | |||
AK214265.1 | - | 1 | 52 | |||
BC002242.1 | - | 3 | 297 | 3492262 |