Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026983.2 | NP_081259.1 | 2 | 215 | ||
GenBank mRNA | AK007684.1 | - | 2 | 216 | ||
AK007800.1 | - | 2 | 136 | |||
AK131830.1 | - | 2 | 216 | |||
BC037376.1 | - | 1 | 51 | |||
BC104406.1 | - | 2 | 203 | 40050532 | ||
BC104407.2 | - | 2 | 203 | 40050533 |