Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093752.1 | NP_001087221.1 | 8 | 863 | ||
NM_001093753.1 | NP_001087222.1 | 7 | 845 | |||
GenBank mRNA | AK011472.1 | - | 8 | 801 | ||
AK029394.1 | - | 7 | 808 | |||
AK081511.1 | - | 5 | 670 | |||
AK146719.1 | - | 7 | 817 | |||
BC002111.1 | - | 7 | 816 | |||
BC021809.1 | - | 7 | 800 | |||
BC037053.1 | - | 8 | 786 | |||
BC038928.1 | - | 8 | 822 | |||
BC055763.1 | - | 9 | 750 | |||
BC069945.1 | - | 7 | 795 | |||
BC138145.1 | - | 5 | 598 | 8861164 | ||
BC145162.1 | - | 5 | 598 | 9053713 |