Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdspl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133710.3 | NP_598471.3 | 6 | 781 | ||
GenBank mRNA | AA239542.1 | - | 6 | 472 | ||
AJ344340.1 | - | 6 | 863 | |||
AK013115.1 | - | 6 | 777 | |||
AK142857.1 | - | 1 | 564 | |||
BC094289.1 | - | 6 | 466 | 4986338 |