Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162973.1 | NP_001156445.1 | 5 | 711 | ||
NM_001162974.1 | NP_001156446.1 | 3 | 839 | |||
NM_027053.1 | NP_081329.1 | 6 | 774 | |||
GenBank mRNA | AK005758.1 | - | 6 | 725 | ||
BC025128.1 | - | 3 | 890 | 4225298 | ||
EU627089.1 | - | 4 | 575 |
Gene Symbol: Lamtor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080275.1 | - | 1269 |