Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198095.2 | NP_932763.1 | 4 | 447 | ||
GenBank mRNA | AK009361.1 | - | 4 | 247 | ||
AK154399.1 | - | 4 | 440 | |||
BC027328.1 | - | 4 | 481 | 4009434 | ||
BC056638.1 | - | 4 | 454 | 5065707 | ||
BC087949.1 | - | 4 | 439 | 6768735 |