Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027149.2 | NP_081425.2 | 1 | 98 | ||
GenBank mRNA | AK009711.1 | - | 1 | 128 | ||
AK052481.1 | - | 1 | 46 | |||
AK136417.1 | - | 1 | 58 | |||
AK145389.1 | - | 1 | 21 | |||
BC026761.1 | - | 1 | 38 | 4221717 | ||
BC027497.1 | - | 34 | ||||
BC064773.1 | - | 1 | 55 |
Gene Symbol: 4930421J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015171.1 | - | 1 | 303 | ||
AK076722.1 | - | 1 | 304 |