Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197991.2 | NP_932108.2 | 2 | 310 | ||
GenBank mRNA | AK150499.1 | - | 2 | 215 | ||
AK160274.1 | - | 2 | 225 | |||
AK169010.1 | - | 2 | 261 | |||
AK171657.1 | - | 2 | 224 | |||
AY761096.1 | - | 2 | 203 | |||
AY761098.1 | - | 2 | 203 | |||
BC020179.1 | - | 2 | 205 | 4913500 |