Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027182.2 | NP_081458.1 | 2 | 397 | ||
GenBank mRNA | AK010336.1 | - | 2 | 359 | ||
AK142463.1 | - | 370 | ||||
AK146877.1 | - | 2 | 398 | |||
AK151568.1 | - | 2 | 333 | |||
BC064315.1 | - | 2 | 320 | |||
BC126946.1 | - | 2 | 296 | 40090292 |