Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027194.3 | NP_081470.2 | 4 | 786 | ||
GenBank mRNA | AK010545.1 | - | 4 | 718 | ||
AK045768.1 | - | 4 | 745 | |||
AK080854.1 | - | 4 | 713 | |||
AK083654.1 | - | 4 | 707 | |||
AK085975.1 | - | 4 | 738 | |||
AK086015.1 | - | 4 | 722 | |||
AK086059.1 | - | 4 | 741 | |||
AK088246.1 | - | 4 | 1251 | |||
AK149407.1 | - | 4 | 715 | |||
AK160039.1 | - | 4 | 723 | |||
AK166012.1 | - | 4 | 739 | |||
AK210124.1 | - | 1 | 133 | |||
BC026789.1 | - | 4 | 713 | 4188168 | ||
CO044843.1 | - | 1 | 509 |