Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145954.1 | NP_666066.1 | 13 | 1765 | ||
GenBank mRNA | AK030673.1 | - | 13 | 1758 | ||
AK144967.1 | - | 1764 | ||||
AK144995.1 | - | 13 | 1772 | |||
AK151691.1 | - | 13 | 1765 | |||
AK153129.1 | - | 6 | 672 | |||
AK161198.1 | - | 13 | 1803 | |||
AK164374.1 | - | 13 | 1811 | |||
AK169346.1 | - | 2116 | ||||
AK170276.1 | - | 13 | 1804 | |||
AK212056.1 | - | 2 | 208 | |||
AK219145.1 | - | 3 | 318 | |||
AK220200.1 | - | 13 | 1741 | |||
BC013548.1 | - | 13 | 1765 | 4222058 |