Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199008.2 | NP_950173.1 | 4 | 696 | ||
GenBank mRNA | AK008096.1 | - | 4 | 710 | ||
AK143937.1 | - | 4 | 696 | |||
AK172067.1 | - | 4 | 700 | |||
BC061233.1 | - | 4 | 707 | 6585499 |
Gene Symbol: Tom1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083630.1 | - | 3328 |