Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027222.3 | NP_081498.2 | 4 | 528 | ||
GenBank mRNA | AA866731.1 | - | 2 | 154 | ||
AK008016.1 | - | 4 | 529 | |||
AK088094.1 | - | 4 | 548 | |||
BC030674.1 | - | 3 | 310 | |||
BC064044.1 | - | 4 | 515 | 875322 | ||
GQ477351.1 | - | 4 | 494 |