Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023537.5 | NP_076026.1 | 4 | 580 | ||
GenBank mRNA | AB232588.1 | - | 3 | 431 | ||
AF312036.1 | - | 3 | 431 | |||
AK012165.1 | - | 4 | 540 | |||
AK082959.1 | - | 4 | 580 | |||
BC051918.1 | - | 4 | 485 | 5691723 | ||
BC057173.1 | - | 3 | 449 | 4989309 |