Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023538.2 | NP_076027.1 | 8, 9 | 569 | ||
GenBank mRNA | AJ401619.1 | - | 8, 9 | 542 | ||
AK076224.1 | - | 7, 8 | 532 | |||
AY772469.1 | - | 7, 8 | 510 | |||
BC019145.1 | - | 8, 9 | 563 | 5051535 | ||
BC093525.1 | - | 8, 9 | 539 | 5151866 |