Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apitd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027263.2 | NP_081539.1 | 2 | 164 | ||
GenBank mRNA | AK011725.1 | - | 2 | 113 | ||
AK012995.1 | - | 2 | 146 | |||
AK013038.1 | - | 2 | 146 | |||
BC048521.1 | - | 2 | 141 | 6741262 | ||
BC138926.1 | - | 2 | 105 | 8861948 | ||
BC138927.1 | - | 2 | 105 | 8861949 | ||
DV656255.1 | - | 2 | 164 |