Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tunar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012713.1 | - | 2 | 374 | ||
AK045952.1 | - | 3 | 480 | |||
BC059025.1 | - | 3 | 726 |