Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122732.1 | NP_001116204.1 | 1 | 287 | ||
NM_027029.2 | NP_081305.1 | 1 | 287 | |||
NM_027300.3 | NP_081576.2 | 1 | 287 | |||
NM_028647.4 | NP_082923.1 | 1 | 287 | |||
GenBank mRNA | AF399971.1 | - | 1 | 286 | ||
AK005880.1 | - | 1 | 286 | |||
AK006491.1 | - | 1 | 277 | |||
AK015193.1 | - | 1 | 285 | |||
AK131709.1 | - | 1 | 286 | |||
BC048516.1 | - | 1 | 286 | |||
BC061025.1 | - | 1 | 268 | 6772738 | ||
BU936341.1 | - | 1 | 271 | |||
BU938516.1 | - | 1 | 289 | |||
EF546784.2 | - | 1 | 279 | |||
EU259321.1 | - | 1 | 279 |