Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133719.2 | NP_598480.1 | 2 | 540 | ||
GenBank mRNA | AK007688.1 | - | 2 | 480 | ||
AK087402.1 | - | 2 | 541 | |||
BC008695.1 | - | 2 | 418 | |||
BC037181.1 | - | 2 | 467 | |||
BC132177.1 | - | 3 | 542 | 40130454 | ||
DQ133462.1 | - | 2 | 463 | |||
DQ133463.1 | - | 2 | 171 | |||
DQ133464.1 | - | 2 | 171 |