Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027323.2 | NP_081599.2 | 3 | 227 | ||
GenBank mRNA | AK012640.1 | - | 3 | 192 | ||
BC107023.2 | - | 3 | 205 | |||
BC125007.1 | - | 3 | 210 | |||
BY291840.1 | - | 3 | 229 |