Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027338.1 | NP_081614.1 | 4 | 316 | ||
GenBank mRNA | AK008946.1 | - | 4 | 354 | ||
AK150754.1 | - | 4 | 337 | |||
AK151035.1 | - | 4 | 337 | |||
AK156241.1 | - | 330 | ||||
AK161865.1 | - | 368 | ||||
BC010811.1 | - | 4 | 316 | 4210911 |