Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122604.1 | NP_001116076.1 | 8, 9 | 848 | ||
NM_001122605.1 | NP_001116077.1 | 8, 9 | 845 | |||
NM_133188.2 | NP_573451.2 | 8, 9 | 845 | |||
GenBank mRNA | AF225910.1 | - | 8, 9 | 739 | ||
AK148023.1 | - | 8, 9 | 723 | |||
AK152314.1 | - | 8, 9 | 815 | |||
AK152345.1 | - | 8, 9 | 815 | |||
BC049355.1 | - | 8, 9 | 765 | 6337388 | ||
BC058775.1 | - | 8, 9 | 759 |