Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197995.2 | NP_932112.2 | 3, 4 | 415 | ||
GenBank mRNA | AK038349.1 | - | 3, 4 | 287 | ||
AK140077.1 | - | 2, 3 | 419 | |||
BC139260.1 | - | 3, 4 | 286 | 8862282 | ||
BC139262.1 | - | 3, 4 | 286 | 8862284 |