Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178379.3 | NP_848466.1 | 7 | 1314 | ||
GenBank mRNA | AK010385.1 | - | 7 | 1297 | ||
AK149999.1 | - | 7 | 1304 | |||
AK154668.1 | - | 7 | 1313 | |||
AK171661.1 | - | 7 | 1304 | |||
BC018301.1 | - | 7 | 1265 | |||
BC038046.1 | - | 7 | 1267 | 4206826 |