Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178379.3 | NP_848466.1 | 6 | 927 | ||
GenBank mRNA | AK010385.1 | - | 6 | 910 | ||
AK149999.1 | - | 6 | 917 | |||
AK154668.1 | - | 6 | 926 | |||
AK171661.1 | - | 6 | 917 | |||
AK178210.1 | - | 3 | 249 | |||
AK184676.1 | - | 3 | 351 | |||
AK186700.1 | - | 3 | 249 | |||
BC018301.1 | - | 6 | 878 | |||
BC038046.1 | - | 6 | 880 | 4206826 |