Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027411.2 | NP_081687.2 | 5 | 829 | ||
GenBank mRNA | AK011138.1 | - | 5 | 801 | ||
AK139553.1 | - | 5 | 705 | |||
BC006674.1 | - | 5 | 812 | 3499487 | ||
BC025900.1 | - | 2 | 747 |