Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027412.3 | NP_081688.2 | 2 | 339 | ||
GenBank mRNA | AK008753.1 | - | 1 | 670 | ||
AK169288.1 | - | 2 | 340 | |||
AK169296.1 | - | 2 | 340 | |||
BC035304.1 | - | 1 | 250 | |||
BC043061.1 | - | 1 | 651 | 5704924 | ||
BC058967.1 | - | 1 | 651 | 5704924 | ||
BC063773.1 | - | 1 | 305 | 30147949 |