Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027412.3 | NP_081688.2 | 3 | 618 | ||
GenBank mRNA | AK008753.1 | - | 2 | 949 | ||
AK018145.1 | - | 3 | 658 | |||
AK169288.1 | - | 3 | 619 | |||
AK169296.1 | - | 3 | 619 | |||
BC035304.1 | - | 2 | 529 | |||
BC043061.1 | - | 2 | 930 | 5704924 | ||
BC058967.1 | - | 2 | 930 | 5704924 |