Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027432.3 | NP_081708.1 | 4 | 555 | ||
GenBank mRNA | AK012014.1 | - | 4 | 546 | ||
AK032897.1 | - | 2 | 94 | |||
AK048488.1 | - | 4 | 578 | |||
AK050391.1 | - | 4 | 555 | |||
AK078856.1 | - | 5 | 514 | |||
AK131608.1 | - | 3 | 466 | |||
AK161812.1 | - | 4 | 530 | |||
AK163839.1 | - | 550 | ||||
AK168006.1 | - | 4 | 557 | |||
AK168752.1 | - | 4 | 577 | |||
AK169128.1 | - | 4 | 556 | |||
BC005755.1 | - | 4 | 510 | 2654151 |