Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001321.3 | NP_001001321.2 | 6, 7 | 479 | ||
GenBank mRNA | AB117931.1 | - | 6, 7 | 460 | ||
AB117932.1 | - | 6, 7 | 460 | |||
AK154257.1 | - | 6, 7 | 505 | |||
AK159148.1 | - | 6, 7 | 480 | |||
BC046402.1 | - | 19 | ||||
BC107210.1 | - | 5, 6 | 426 | |||
BC107211.1 | - | 6, 7 | 459 | 40051417 |