Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081242.2 | NP_001074711.2 | 40 | 5317 | ||
GenBank mRNA | AB093231.1 | - | 14 | 1859 | ||
AF467081.1 | - | 38 | 5046 | |||
AK029828.1 | - | 15 | 2010 | |||
BC033288.1 | - | 4 | 480 | |||
BC158057.1 | - | 40 | 5288 | 9007357 |